Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCORL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548848

Catalog No. PA548848

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May act as transcription activator that binds DNA elements with the sequence 5 ft.-CCCTATCGATCGATCTCTACCT-3 ft.. May play a role in spermatogenesis.
Specifications

Specifications

LCORL
Polyclonal
PBS with 0.09% sodium azide
Q8N3X6
Ligand-dependent nuclear receptor corepressor-like protein, LCoR-like protein, LCORL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
254251
Western Blot
Unconjugated
LCORL
Liquid
LCORL
KLH conjugated synthetic peptide between 306-334 amino acids from the Central region of human LCORL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit