Learn More
Teledyne SSI LD Class Pump
The versatile LD Class consists of dual-headed, positive displacement piston pumps, delivering performance for analytical, flash, and small-scale preparative chromatography applications. 
$6780.00
Specifications
For Use With (Application) mL/Min
View More Specs
Products 2
Catalog Number Mfr. No. Flow Rate Pressure Material Includes Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Pressure Material Includes Price Quantity & Availability  
06-657-014 View Documents Teledyne SSI
LD012SFT1A
6000 PSI Stainless Steel Stainless Steel Energizer Standard Piston Seal Each for $6,780.00
Add to cart
06-657-015 View Documents Teledyne SSI
LD012PFT3A
12 mL/min 5000 PSI Polyether Ether Ketone (PEEK) Flouropolymer Energizer Standard Piston Seal Each for $6,780.00
Add to cart
Description
Description
  • Self-Flush and Piston Wash
  • Pulse Dampener or Pressure & Leak Sensor
  • Inlet Tubing: 1/8in. OD Flexible
  • Outlet Tubing: 1/16in. OD
  • The pumps are designed for high flow rate accuracy and low pulsation required for Analytical Liquid Chromatography (HPLC). Standard fluid path materials are Stainless Steel and PEEK. With 12 mL/min version, reaching pressures up to 6,000 psi, these pumps will meet the performance requirements of demanding HPLC needs.
  • The high performance linear cams, along with automatic pressure compensation, provide precise flow rates for very accurate high pressure and low pressure gradients. Standard features include an integrated Prime-Purge Valve, Pulse Dampener, interactive keypad control, and complete PC control and status through RS-232 and Micro USB 2.0 ports. The LD Class provides the precise flow and low pulsation for the most challenging separations.
Specifications
Specifications
mL/Min