Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDB3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$152.22 - $436.00
Specifications
Antigen | LDB3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15499820
|
Novus Biologicals
NBP15499820UL |
20 μL |
Each for $152.22
|
|
NBP154998
|
Novus Biologicals
NBP154998 |
100 μL |
Each for $436.00
|
|
Description
LDB3 Polyclonal specifically detects LDB3 in Human, Mouse samples. It is validated for Western Blot.Specifications
LDB3 | |
Polyclonal | |
Purified | |
RUO | |
O75112-5 | |
11155 | |
Synthetic peptides corresponding to LDB3(LIM domain binding 3) The peptide sequence was selected from the N terminal of LDB3. Peptide sequence PVIPHQKDPALDTNGSLVAPSPSPEARASPGTPGTPELRPTFSPAFSRPS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CMD1C, CYPHER, KIAA01613, KIAA0613FLJ35865, LDB3Z1, LDB3Z4, LIM domain binding 3, LIM domain-binding protein 3, ORACLE, PDLIM6, PDZ and LIM domain 6, Protein cypher, ZASPLVNC3, Z-band alternatively spliced PDZ-motif protein | |
LDB3 | |
IgG | |
Protein A purified | |
36 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title