Learn More
Selleck Chemical LLC
Sonidegib (NVP-LDE225) 50mg 956697-53-3 Erismodegib
Manufacturer: Selleck Chemical LLC S215150MG
Sonidegib (Erismodegib, NVP-LDE225) is a Smoothened (Smo) antagonist, inhibiting Hedgehog (Hh) signaling with IC50 of 1.3 nM (mouse) and 2.5 nM (human) in cell-free assays, respectively. Phase 3.