Learn More
Selleck Chemical LLCSupplier Diversity Partner Sonidegib (NVP-LDE225) 10mg 956697-53-3 Erismodegib  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S215110MG
Sonidegib (Erismodegib, NVP-LDE225) is a Smoothened (Smo) antagonist, inhibiting Hedgehog (Hh) signaling with IC50 of 1.3 nM (mouse) and 2.5 nM (human) in cell-free assays, respectively. Phase 3.
Catalog No. 50-643-9