Learn More
Thermo Scientific™ LDH Reagents
Determine in vitro quantity of LDH in human serum in vitro with Thermo Scientific™ LDH Reagents.
Catalog No. TR20015
Add to cart
Description
Description
Determine in vitro quantity of LDH in human serum in vitro with Thermo Scientific™ LDH Reagents.
Lactate dehydrogenase (LDH) is concentrated in the liver, heart, kidney, skeletal muscle and erythrocytes. Increases are observed after myocardial infarction and in liver disease, untreated pernicious anemia, renal disease, malignancy and progressive muscular dystrophy.
  • At least five days reconstituted stability
  • For use in manual or automated systems
Specifications
Specifications
CE Marked
Reagent
Infinity Triglyceride 2 x 125mL
Up to 10mmol/L (885mg/dL)
18 months at 2-8°C
Plasma, Serum, Serum or Plasma
Yes
LDH
Endpoint
SDS
Documents
Accessories