Learn More
Aviva Systems Biology Corp LDHA Antibody - N-terminal region (ARP54776_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LDHA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LDHA. Alternate names: LDH-M, LDH1, PIG19, LDHM, GSD11ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61308