Learn More
Aviva Systems Biology Corp LDHB Antibody - C-terminal region (ARP48210_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LDHB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human LDHB. Alternate names: LDH-H, TRG-5, LDH-B, LDHBDApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59760