Learn More
Abnova™ LDHC 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00003948T02
Catalog No. 89-950-509
Add to cart
Specifications
Specifications
293T
NM_002301.2
pCMV-LDHC full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LDHC 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.3kDa
Transient overexpression cell lysate was tested with anti-LDHC antibody (Mfr. No. H00003948-D01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human