Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova LDHC 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer: Abnova H00003948T01
Specifications
293T | |
NM_002301.2 | |
pCMV-LDHC full-length | |
100 uL | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Western Blot |
LDHC 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 36.63kDa | |
Transient overexpression cell lysate was tested with anti-LDHC antibody (Mfr. No. H00003948-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title