Learn More
Athens Research And Technology IncSupplier Diversity Partner LDL LIPOPROTEINS HU 5MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 12161204125MG
Lipoproteins, Low Density, Human Plasma: 5 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 2,300,000. Native; Prepared and purified from fresh, non-frozen human plasma in the Athens Research & Technology laboratory. Lipoproteins, Molecular Biology Reagent
Catalog No. 50-489-866