Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDL, MDA treated Mouse anti-Human, Alexa Fluor™ 532, Clone: 4C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223597AF532

Catalog No. NP22397A32


Add to cart

Description

Description

LDL, MDA treated Monoclonal antibody specifically detects LDL, MDA treated in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

LDL, MDA treated
4C7
Western Blot, ELISA
Conjugated, Purified
Mouse
IgG2b Kappa
0.1 mL
Primary
It reacts with MDA-treated LDL but not with native LDL
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Low density lipoprotein, MDA oxidized LDL, MDA oxidized low density lipoprotein
Low-density lipoprotein purified from human plasma and treated with malonyldialdehyde (MDA) and adsorbed onto aluminum hydroxide gel
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit