Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR 293T Cell Overexpression Lysate (Denatured), Abnova
Denatured human lysate for western blotting
Manufacturer: Abnova Corporation H00003949T01
Description
- Transfected cell line: 293T
- Plasmid: pCMV-LDLR full-length
- Host: Human
- Theoretical molecular weight: 94.6kDa
- Storage buffer: 1X sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western blotting
Specifications
Western Blot | |
LDLR 293T Cell Transient Overexpression Lysate(Denatured) | |
94.6 | |
pCMV-LDLR full-length | |
100 μL | |
Human |
293T | |
BC014514 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-LDLR antibody (H00003949-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title