Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Mouse anti-Human, Alexa Fluor™ 350, Clone: 11A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222035AF350

Catalog No. NP22203A30


Add to cart

Description

Description

LDLR Monoclonal antibody specifically detects LDLR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

LDLR
11A2
Western Blot, Flow Cytometry, ELISA
Conjugated, Purified
LDLR
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant LDL R. Mouse anti human LDL R P01130.
Protein G purified
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
3949.0
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit