Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Antibody (11A2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222035AF750
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
LDLR | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
LDLR | |
Human recombinant LDLR | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
11A2 | |
Western Blot, Flow Cytometry, ELISA | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Mouse | |
Protein G purified | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
3949.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction