Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Mouse anti-Human, PE, Clone: C7, BD Pharmingen™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565653

Catalog No. BDB565653


Add to cart

Description

Description

LDLR Monoclonal antibody specifically detects LDLR in Cow , Human samples. It is validated for Flow cytometry.
Specifications

Specifications

LDLR
C7 (also known as IgG-C7)
PE
Purified
Murine
Mouse IgG2b, κ
RUO
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
low-density lipoprotein receptor; LDL receptor; FH; FHC; LDLCQ2
Cow adrenal LDL receptor
50μg
Cell Biology
Primary
Bovine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.