Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Rabbit anti-Human, Alexa Fluor 405, Clone: 301, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289351AF405
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.Specifications
LDLR | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
301 | |
Flow Cytometry | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human LDLR (NP_000518.1; Met1-Arg788). | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
3949 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title