Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Rabbit anti-Human, Alexa Fluor 594, Clone: 301, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289351AF594

Catalog No. NB066376


Add to cart

Description

Description

LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

LDLR
301
Flow Cytometry
Purified
Rabbit
IgG
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
3949
Flow Cytometry
Alexa Fluor 594
50mM Sodium Borate
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
This antibody was obtained from a rabbit immunized with purified, recombinant Human LDLR (NP_000518.1; Met1-Arg788).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit