Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Rabbit anti-Mouse, Alexa Fluor 532, Clone: 32, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290523AF532

Catalog No. NB039844


Add to cart

Description

Description

LDLR Monoclonal antibody specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

LDLR
32
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
3949
ELISA, Flow Cytometry, Immunofluorescence
Alexa Fluor 532
50mM Sodium Borate
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Recombinant Mouse LDLR protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit