Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLRAP1 Mouse anti-Human, Clone: OTI7E11, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503854

Catalog No. 50-167-4640


Add to cart

Description

Description

The protein encoded by this gene is a cytosolic protein which contains a phosphotyrosine binding (PTD) domain. The PTD domain has been found to interact with the cytoplasmic tail of the LDL receptor. Mutations in this gene lead to LDL receptor malfunction and cause the disorder autosomal recessive hypercholesterolaemia.
Specifications

Specifications

LDLRAP1
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5SW96
ARH, ARH1, ARH2, FHCB1, FHCB2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26119
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7E11
Unconjugated
LDLRAP1
Liquid
LDLRAP1
Full length human recombit protein of human LDLRAP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit