Learn More
WASP Project LDM WASP Extruder XL Products for Science Education
Professional LDM / ceramic extruder for any low density fluid material.
Manufacturer:  WASP Project PS7001
Catalog No. S81205
Add to cart
Description
Description
  • Equipped with a screw system capable of regulating the output of material and allowing rapid flow and great retraction control
  • Only available on the 40100 or the 3MT
  • New ceramic extruder for the Delta WASP 40100 Clay which allows fast, precise, large prints with hard dough.
  • Degasser extruder innovation avoids air bubbles in the dough
  • Extrusion control with possibility of retraction
  • High torque motor for molding hard dough - nozzles up to 8 mm in diameter.
  • Allows the printing of harder doughs and therefore faster printing by exploiting the entire print volume of the Delta WASP 40100 Clay.
  • Precisely manage the flow of material at the exit up to the use of retraction to stop the deposition
Specifications
Specifications
STEM Project
Low density materials, clay, porcelain, stoneware, earthenware