Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDN 212320, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Increases expression of glutamate transporter EAAT2 in PA-EAAT2 cells. Displays neuroprotective activity in vivo. Shown to improve learning and memory, restore synaptic integrity and reduce amyloid plaque burden in APPSw/Ind mice.
Specifications
Quantity | 10 mg |
Formula Weight | 293.39 |
Chemical Name or Material | LDN 212320 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction