LDN 57444, Tocris™

Ubiquitin C-terminal hydrolase-L1 (UCH-L1) inhibitor

Manufacturer: R&D Systems 3998/10

Quantity 10mg

 View more versions of this product

Catalog No. 39-981-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C17H11Cl3N2O3
    Formula Weight 397.64
    Synonym 5-Chloro-1-[(2, 5-dichlorophenyl)methyl]-1H-indole-2, 3-dione 3-(O-acetyloxime)
    Chemical Name or Material LDN 57444

    Specifications

    Formula Weight 397.64
    Quantity 10mg
    Assay Percent Range >98%
    CAS 668467-91-2
    Molecular Formula C17H11Cl3N2O3
    Synonym 5-Chloro-1-[(2, 5-dichlorophenyl)methyl]-1H-indole-2, 3-dione 3-(O-acetyloxime)
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material LDN 57444

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit