Learn More
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
Click to view available options
Closure Size:
24-410 mm
28-410 mm
38-400 mm
48-400 mm
53-400 mm
63-400 mm
100-400 mm
Liner Material:
LDPE Foam
No. per Case:
70
90
200
250
500
Use these LDPE foam-lined caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ LDPE Foam-lined Polypropylene White Caps are for general-purpose HDPE bulk separate containers. The same high quality caps used on our Thermo Scientific certified HDPE environmental sample containers. Bottles sold separately (013-series and 012-series). Unprocessed and uncertified.
  • LDPE foam-lined white polypropylene cap
  • Bottles sold separately: 013-series and 012-series
  • Recommended for general purpose sampling where cleaning and certification is not required, or ready for your own cleaning and certification procedures
  • Economically bulk packed
Includes: Cap only
Closure Included Yes
Closure Liner LDPE Foam
Shape Round
Certifications/Compliance Manufactured under an ISO certified quality management system
Closure Size 63-400 mm
Certification Parameters None
Color White
Description White PP Cap
Material Polypropylene
No. per Case 90
Septa N/A
Type Closure
Level Unprocessed
Liner Material LDPE Foam
Show More Show Less