Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, Trihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
Packaging:
Poly Bottle

Chemical Identifiers

6080-56-4
C4H12O7Pb
379.333
MCEUZMYFCCOOQO-UHFFFAOYSA-L
22456
CHEBI:33112
lead(2+);diacetate;trihydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 379.33
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 2.55kg
Quantity 500g
Chemical Name or Material Lead Acetate, Trihydrate
Grade Baker Analyzed™ ACS Reagent