Lead Acetate, Trihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 227105

Packaging Poly Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. 02-003-337

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6080-56-4
    Molecular Formula (CH3COO)2Pb·3H2O
    Formula Weight 379.33
    Chemical Name or Material Lead Acetate, Trihydrate
    Grade Baker Analyzed™ ACS Reagent

    Specifications

    Formula Weight 379.33
    Molecular Formula (CH3COO)2Pb·3H2O
    Packaging Poly Bottle
    Physical Form Granular
    Specific Gravity 1L = 2.55kg
    CAS 6080-56-4
    Quantity 2.5kg
    Chemical Name or Material Lead Acetate, Trihydrate
    Grade Baker Analyzed™ ACS Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit