Alfa Aesar™ Lead Antimony shot, 2mm (0.08in)

Manufacturer: Alfa Aesar™ 1022436

Quantity 500g

 View more versions of this product

Catalog No. AA1022436

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00134050
    UN Number UN3077
    Chemical Name or Material Lead Antimony shot, 2mm (0.08 in.)

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Shot
    MDL Number MFCD00134050
    UN Number UN3077
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332-H201
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Explosive
    mass explosion hazard.
    TSCA TSCA
    Odor Odorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 500g
    Chemical Name or Material Lead Antimony shot, 2mm (0.08 in.)

    GHS H Statement
    H360-H373-H302-H332-H201
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Explosive
    mass explosion hazard.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit