Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate), 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
21319-43-7
C22 H38 O4 Pb
MFCD00070468
21319-43-7
C22 H38 O4 Pb
MFCD00070468
Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point 128°C to 131°C
Quantity 5g
Color White
Assay Percent Range 99+%
Formula Weight 573.74
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate), 99+%

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertil

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger