Lead Foil, 0.1mm (0.004 in.) thick, 99.9% (Metals basis), Alfa Aesar™

Form: Approx.25.5g/15 x 15cm

Manufacturer: Alfa Aesar 42708-VA

Quantity 15 x 15cm

 View more versions of this product

Catalog No. AA42708VA

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7439-92-1
    Physical Form Foil, 0.1mm (0.004 in.) thick
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00134050
    Chemical Name or Material Lead

    Specifications

    Quantity 15 x 15cm
    Color Gray
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Density 11.34g/cm3 at 20°C
    EINECS Number 231-100-4
    Physical Form Foil, 0.1mm (0.004 in.) thick
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00134050
    CAS 7439-92-1
    Odor Odorless
    Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Appearance Gray foil
    Chemical Name or Material Lead

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit