Lead Foil, 1.19mm (0.0469 in.) thick, 99.8% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 12604VA

Quantity 15 x 15cm

 View more versions of this product

Catalog No. AA12604VA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-92-1
    Physical Form Foil, 1.19mm (0.0469 in.) thick
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00134050
    Chemical Name or Material Lead

    Specifications

    Quantity 15 x 15cm
    Color Gray
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    EINECS Number 231-100-4
    Physical Form Foil, 1.19mm (0.0469 in.) thick
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00134050
    CAS 7439-92-1
    Odor Odorless
    Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Appearance Gray foil
    Chemical Name or Material Lead

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit