Learn More
SPS Medical LEAD FREE AUTOCLVE TAPE 1'X60Y DFS Item
Manufacturer:  SPS Medical LF2036
Indicator tapes provide immediate identification of processed items while also securing sterilization packs. Color change indicator verifies exposure to sterilization process. Tapes are individually packaged in 60 yard rolls and available in multiple widths. Blue steam tape has been specially designed with aggressive adhesive for disposable CSR wraps. Tape dispenser can hold up to 2 rolls of 1” tape and has a weighted base.
Catalog No. NC0003837