Alfa Aesar™ Lead(II) acetate trihydrate, 99%

Manufacturer: Alfa Aesar™ A1174630

Quantity 250g

 View more versions of this product

Catalog No. AAA1174630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6080-56-4
    Molecular Formula Pb(OOCCH3)2·3H2O
    Formula Weight 379.33 (325.28 Anhydrous)
    MDL Number MFCD00150023
    UN Number UN1616
    Beilstein 3730298
    Merck Index 14,5397
    Chemical Name or Material Lead (II) acetate trihydrate

    Specifications

    Formula Weight 379.33 (325.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 6080-56-4
    Beilstein 3730298
    RTECS Number OF8050000
    MDL Number MFCD00150023
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
    UN Number UN1616
    Health Hazard 2 GHS H Statement
    H360-H373
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Melting Point 75°C
    Molecular Formula Pb(OOCCH3)2·3H2O
    Merck Index 14,5397
    Quantity 250g
    Chemical Name or Material Lead (II) acetate trihydrate
    Assay 99%

    Description

    Applications
    Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

    Solubility
    Freely soluble in water, alcohol and glycerol.

    Notes
    Decomposes above 200°Air and light sensitive. Incompatible with strong acids and strong oxidizing agents.

    GHS H Statement
    H360-H373
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit