Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) acetate trihydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
6080-56-4
C4H6O4Pb·3H2O
379.33
MFCD00150023
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate
22456
CHEBI:33112
lead(2+);diacetate;trihydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
6080-56-4
C4H6O4Pb·3H2O
379.33
MFCD00150023
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate
22456
CHEBI:33112
lead(2+);diacetate;trihydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
Assay Percent Range 99.0 to 103% (Complexometry)
Linear Formula (CH3CO2)2Pb·3H2O
Merck Index 15, 5452
Solubility Information Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol,freely soluble in glycerol
Formula Weight 379.33
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 50ppm max.
Chloride 5ppm max.
Copper (Cu) 10ppm max.
Iron (Fe) 10ppm max.
Nitrate 50ppm max.
Packaging Plastic bottle
Potassium (K) 50ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
Color White
Melting Point 75.0°C
Quantity 2.5kg
Chemical Name or Material Lead(II) acetate trihydrate, p.a.
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA

GHS Signal Word: Danger

RTECSNumber : OF8050000