Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Lead(Ii) Acetate Trihydrate, ACS Reagent, 99.0 to 103.0%, Honeywell™ Fluka™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Poly Bottle
6080-56-4
C4H12O7Pb
379.333
MFCD00150023
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate
22456
CHEBI:33112
lead(2+);diacetate;trihydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
6080-56-4
C4H12O7Pb
379.333
MFCD00150023
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate
22456
CHEBI:33112
lead(2+);diacetate;trihydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number UN1616
Beilstein 3730298
Formula Weight 379.33g/mol
Percent Purity 99.0 to 103.0%
Grade ACS Reagent
Density 2.550g/cu.cm
Packaging Poly Bottle
Physical State Solid
Boiling Point 204°C
Color Colorless
Melting Point 75°C
Quantity 2.5kg
Chemical Name or Material Lead(Ii) Acetate Trihydrate
Show More Show Less

P201-P273-P308 + P313-P501

H360Df-H373-H410

DOTInformation : 6.1/PGIII

EINECSNumber : 206-104-4

RTECSNumber : OF8050000