Alfa Aesar™ Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 1071906

Quantity 5g

 View more versions of this product

Catalog No. AA1071906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6080-56-4
    Formula Weight 379.33 (325.28 Anhydrous)
    MDL Number MFCD00150023
    UN Number UN1616
    Beilstein 3730298
    Merck Index 14,5397
    Chemical Name or Material Lead(II) acetate trihydrate
    Grade Puratronic™

    Specifications

    Formula Weight 379.33 (325.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 75°C
    Health Hazard 2 GHS H Statement
    H360-H373
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Physical Form Crystalline
    MDL Number MFCD00150023
    UN Number UN1616
    Beilstein 3730298
    Merck Index 14,5397
    Health Hazard 1 Danger
    Density 2.55g/mL
    TSCA TSCA
    CAS 6080-56-4
    Quantity 5g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
    RTECS Number OF8050000
    Chemical Name or Material Lead(II) acetate trihydrate
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H360-H373
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit