Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) bromide, 98+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
10031-22-8
Br2Pb
367.01
MFCD00011156
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
lead ii bromide,lead bromide,lead dibromide,lead bromide pbbr2,pbbr2,lead 2+ bromide,dibromoplumbane,ccris 4220,leadbromide,acmc-1bq2x
24831
dibromolead
Br[Pb]Br
10031-22-8
Br2Pb
367.01
MFCD00011156
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
lead ii bromide,lead bromide,lead dibromide,lead bromide pbbr2,pbbr2,lead 2+ bromide,dibromoplumbane,ccris 4220,leadbromide,acmc-1bq2x
24831
dibromolead
Br[Pb]Br
Assay Percent Range 98% min. (Argentometry)
Linear Formula PbBr2
Fieser 15,182
Merck Index 15, 5458
Solubility Information Solubility in water: 5g/L (20 c). Other solubilities: insoluble in alcohol
Formula Weight 367.01
Physical Form Fine Crystalline Powder
Percent Purity 98+%
Grade Extra Pure
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Density 6.6600g/mL
Insoluble Matter 0.005% max. (in acid)
Loss on Drying 0.1% max.
Packaging Glass bottle
Solubility Solubility in water: 5g/L (20 c). Other solubilities: insoluble in alcohol
Specific Gravity 6.66
Boiling Point 916.0°C
Color White
Melting Point 373.0°C
Quantity 100g
Chemical Name or Material Lead(II) bromide, 98+%
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b

GHS Signal Word: Danger

EINECSNumber : 233-084-4

TSCA : TSCA