Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) bromide, White powder, 99.999% (Metals basis), Alfa Aesar™
Click to view available options
Quantity:
1g
5g
25g
Specifications
Assay Percent Range | 99.999% (Metals basis) |
Solubility Information | Slightly soluble in water. Insoluble in alcohol. Soluble in ammonia and alkali. |
Formula Weight | 367.01 |
Physical Form | Powder, -10 Mesh Beads,Ampouled under argon |
Density | 6.66 |
Odor | Odorless |
Boiling Point | 916°C |
Color | White |
Melting Point | 373°C |
Quantity | 1g |
Show More |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c
H302+H332-H360Df-H373
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 233-084-4
TSCA : Yes
Recommended Storage : Ambient temperatures