Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) carbonate, ACS reagent, ACROS Organics™
Click to view available options
Quantity:
100g
500g
Specifications
Linear Formula | PbCO3 |
Merck Index | 10,1006 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 267.2 |
Grade | ACS Reagent |
Chloride | 0.002% max |
Insoluble Matter | 0.02% max. (in dilute acetic acid) |
Packaging | Plastic bottle |
Solubility | Solubility in water: insoluble. |
Melting Point | 400.0°C |
Show More |
Safety and Handling
GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 209-943-4