Lead(II) carbonate, basic, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4208922

Quantity 100g

 View more versions of this product

Catalog No. AA4208922

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1319-46-6
    Physical Form Powder
    Assay Percent Range 99% (Metal basis)
    Molecular Formula C2H2O8Pb3
    Formula Weight 775.63
    MDL Number MFCD00078155
    Synonym C.I. 77597
    Chemical Name or Material Lead(II) carbonate, Basic

    Specifications

    Formula Weight 775.63
    Quantity 100g
    Color White
    DOT Information DOT Class: 9, Packaging Group: III
    Density 6.14g/cm3 at 20°C
    EINECS Number 215-290-6
    Physical Form Powder
    Assay Percent Range 99% (Metal basis)
    Molecular Formula C2H2O8Pb3
    Appearance White powder
    MDL Number MFCD00078155
    CAS 1319-46-6
    Synonym C.I. 77597
    Health Hazard 1 H301-H332-H360-H373-H400-H410: Toxic if swallowed. Harmful if inhaled. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Lead(II) carbonate, Basic

    H301-H332-H360-H373-H400-H410: Toxic if swallowed. Harmful if inhaled. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit