Alfa Aesar™ Lead(II) carbonate, basic, ACS

Ceramic glaze, exterior paint

Manufacturer: Alfa Aesar™ 3332818

Quantity 50g

 View more versions of this product

Catalog No. AA3332818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1319-46-6
    Linear Formula (PbCO3)2·Pb(OH)2
    Formula Weight 775.63
    MDL Number MFCD00078155
    UN Number UN3077
    Merck Index 14,1005
    Synonym White lead; C.I. 77597
    Chemical Name or Material Lead(II) carbonate, Basic
    Grade ACS Grade

    Specifications

    Formula Weight 775.63
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00078155
    CAS 1319-46-6
    Synonym White lead; C.I. 77597
    Quantity 50g
    UN Number UN3077
    Merck Index 14,1005
    Density 6.14g/mL
    TSCA TSCA
    Physical Form Powder
    Linear Formula (PbCO3)2·Pb(OH)2
    Color White
    Melting Point 400°C (dec.)
    Solubility Information Soluble in acids, insoluble in water or alcohol
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Lead(II) carbonate, Basic
    Grade ACS Grade

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit