Alfa Aesar™ Lead(II) chromate, 98%

Manufacturer: Alfa Aesar™ A1067818

Quantity 50g

 View more versions of this product

Catalog No. AAA1067818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-97-6
    Molecular Formula CrO4Pb
    Linear Formula PbCrO4
    Formula Weight 323.19
    MDL Number MFCD00011158
    UN Number UN3077
    Merck Index 14,5405
    Synonym Chrome Yellow
    Chemical Name or Material Lead (II) chromate

    Specifications

    Formula Weight 323.19
    Health Hazard 3 GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula CrO4Pb
    MDL Number MFCD00011158
    CAS 7758-97-6
    Health Hazard 2 GHS H Statement
    H350-H360-H373-H201
    May cause cancer.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Explosive
    mass explosion hazard.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number GB2975000
    Odor Odorless
    UN Number UN3077
    Merck Index 14,5405
    Synonym Chrome Yellow
    Melting Point 844°C
    Quantity 50g
    Health Hazard 1 Danger
    Linear Formula PbCrO4
    Chemical Name or Material Lead (II) chromate
    Assay 98%

    GHS H Statement
    H350-H360-H373-H201
    May cause cancer.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Explosive
    mass explosion hazard.

    GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit