Lead(II) chromate, ACS, 98%, Alfa Aesar™

Maximum level of impurities: Soluble matter 0.15%, Carbon compounds (as C) 0.01%

Manufacturer: Alfa Aesar 1412522

Quantity 100g

 View more versions of this product

Catalog No. AA1412522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-97-6
    Physical Form Powder
    Assay Percent Range 98%
    Molecular Formula CrO4Pb
    Formula Weight 323.19
    MDL Number MFCD00011158
    Chemical Name or Material Lead(II) chromate, For ACS analysis
    Grade ACS Grade

    Specifications

    Physical Form Powder
    Formula Weight 323.19
    Quantity 100g
    Color Yellow
    DOT Information DOT Class: 9, Packaging Group: III
    Density 6.3g/cm3 at 20°C
    EINECS Number 231-846-0
    Odor Odorless
    Assay Percent Range 98%
    Molecular Formula CrO4Pb
    Appearance Yellow powder
    MDL Number MFCD00011158
    CAS 7758-97-6
    Health Hazard 1 H350-H360-H373-H400-H410: May cause cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Lead(II) chromate, For ACS analysis
    Grade ACS Grade

    H350-H360-H373-H400-H410: May cause cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit