Alfa Aesar™ Lead(II) citrate trihydrate, 99%

Manufacturer: Alfa Aesar™ A1070136

Quantity 500g

 View more versions of this product

Catalog No. AAA1070136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6107-83-1
    Molecular Formula C12H10O14Pb3·3H2O
    Formula Weight 1053.82 (999.77 Anhydrous)
    MDL Number MFCD00150456
    UN Number UN2291
    Synonym Citric acid lead(II) salt trihydrate
    Chemical Name or Material Lead (II) citrate trihydrate

    Specifications

    Formula Weight 1053.82 (999.77 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2291
    TSCA TSCA
    Molecular Formula C12H10O14Pb3·3H2O
    MDL Number MFCD00150456
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    CAS 6107-83-1
    Synonym Citric acid lead(II) salt trihydrate
    Solubility Information Soluble in cold water
    Health Hazard 1 Danger
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Lead (II) citrate trihydrate
    Assay 99%

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit