Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) cyclohexanebutyrate, 94%

Click to view available options
Quantity:
5g
25g
100g
62637-99-4
C20H34O4Pb
545.688
MFCD00036403
CCJZHCZMNGUOCA-UHFFFAOYSA-L
lead cyclohexanebutyrate,lead bis 4-cyclohexylbutyrate,cyclohexanebutanoic acid, lead 2+ salt,cyclohexanebutanoic acid, lead 2+ salt 2:1,lead 2+ bis 4-cyclohexylbutanoate,acmc-20akns,lead ii cyclohexanebutyrate,4-cyclohexylbutanoate; lead 2+,biscyclohexanebutyric acid lead ii salt,$l^ 2-lead 2+ ion bis 4-cyclohexylbutanoate
112896
4-cyclohexylbutanoate;lead(2+)
C1CCC(CC1)CCCC(=O)[O-].C1CCC(CC1)CCCC(=O)[O-].[Pb+2]
62637-99-4
C20H34O4Pb
545.688
MFCD00036403
CCJZHCZMNGUOCA-UHFFFAOYSA-L
lead cyclohexanebutyrate,lead bis 4-cyclohexylbutyrate,cyclohexanebutanoic acid, lead 2+ salt,cyclohexanebutanoic acid, lead 2+ salt 2:1,lead 2+ bis 4-cyclohexylbutanoate,acmc-20akns,lead ii cyclohexanebutyrate,4-cyclohexylbutanoate; lead 2+,biscyclohexanebutyric acid lead ii salt,$l^ 2-lead 2+ ion bis 4-cyclohexylbutanoate
112896
4-cyclohexylbutanoate;lead(2+)
C1CCC(CC1)CCCC(=O)[O-].C1CCC(CC1)CCCC(=O)[O-].[Pb+2]
Quantity 25g
UN Number UN2291
Formula Weight 545.68
Percent Purity 94%
Chemical Name or Material Lead(II) cyclohexanebutyrate

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

EINECSNumber : 263-663-7

TSCA : Yes

Recommended Storage : Ambient temperatures