Alfa Aesar™ Lead(II) fluoride, Puratronic™, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 1072322

Quantity 100g

 View more versions of this product

Catalog No. AA1072322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-46-2
    Formula Weight 245.2
    MDL Number MFCD00011162
    UN Number UN2291
    Merck Index 14,5408
    Chemical Name or Material Lead(II) fluoride
    Grade Puratronic™

    Specifications

    Formula Weight 245.2
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011162
    Quantity 100g
    Health Hazard 1 Danger
    Density 8.445g/mL
    Physical Form Powder
    UN Number UN2291
    Boiling Point 1290°C
    TSCA TSCA
    Merck Index 14,5408
    Odor Odorless
    CAS 7783-46-2
    Melting Point 855°C
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    Chemical Name or Material Lead(II) fluoride
    Assay 99.997%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit