Alfa Aesar™ Lead(II) hexacyanoferrate(II)

Manufacturer: Alfa Aesar™ A1017436

Quantity 500g

 View more versions of this product

Catalog No. AAA1017436

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 14402-61-0
    Molecular Formula C6FeN6Pb2
    Linear Formula Pb2Fe(CN)6
    Formula Weight 626.35
    MDL Number MFCD00168085
    UN Number UN2291
    Synonym Lead(II) ferrocyanide
    Chemical Name or Material Lead (II) hexacyanoferrate (II)

    Specifications

    Formula Weight 626.35
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 14402-61-0
    UN Number UN2291
    Synonym Lead(II) ferrocyanide
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H331-H360-H373-H302
    Toxic if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Molecular Formula C6FeN6Pb2
    Linear Formula Pb2Fe(CN)6
    MDL Number MFCD00168085
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    Chemical Name or Material Lead (II) hexacyanoferrate (II)

    GHS H Statement
    H331-H360-H373-H302
    Toxic if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit