Alfa Aesar™ Lead(II) metaborate monohydrate

Manufacturer: Alfa Aesar™ 3933914

Quantity 25g

 View more versions of this product

Catalog No. AA3933914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10214-39-8
    Linear Formula PbB2O4·H2O
    Formula Weight 310.81 (292.81 Anhydrous)
    MDL Number MFCD00049504
    UN Number UN2291
    Synonym Lead(II) borate
    Chemical Name or Material Lead(II) metaborate monohydrate

    Specifications

    Formula Weight 310.81 (292.81 Anhydrous)
    Melting Point 160°C (in -H2O)
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Odor Odorless
    MDL Number MFCD00049504
    UN Number UN2291
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    Physical Form Powder
    Quantity 25g
    Solubility Information Insoluble in water and alkali; Soluble in acid
    Health Hazard 1 Danger
    Density 5.598g/mL
    TSCA TSCA
    Linear Formula PbB2O4·H2O
    CAS 10214-39-8
    Synonym Lead(II) borate
    Chemical Name or Material Lead(II) metaborate monohydrate

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit