Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Melting Point | 470.0°C |
Color | White |
Physical Form | Crystalline Solid |
Quantity | 10 g |
Assay Percent Range | 99.999% |
Linear Formula | Pb(NO3)2 |
Merck Index | 15, 5469 |
Solubility Information | Solubility in water: 343g/L. |
Formula Weight | 331.2 |
Percent Purity | 99.999% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume |
EINECSNumber | 233-245-9 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction