Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) nitrate, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
10099-74-8
N2O6Pb
331.2
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate,lead nitrate,lead ii nitrate,plumbous nitrate,lead 2+ nitrate,lead nitrate pb no3 2,nitrate de plomb french,lead ii nitrate 1:2,nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
10099-74-8
N2O6Pb
331.2
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate,lead nitrate,lead ii nitrate,plumbous nitrate,lead 2+ nitrate,lead nitrate pb no3 2,nitrate de plomb french,lead ii nitrate 1:2,nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula Pb(NO3)2
Merck Index 15, 5469
Solubility Information Solubility in water: 343g/L.
Formula Weight 331.2
Physical Form Crystalline Solid
Percent Purity 99.999%
Grade Trace Metal Basis
Density 4.5300g/mL
Packaging Glass bottle
Solubility Solubility in water: 343g/L.
Specific Gravity 4.53
Total Trace Metal Impurities 0.001% max.
Color White
Melting Point 470.0°C
Quantity 50g
Chemical Name or Material Lead(II) nitrate
Show More Show Less

GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume

GHS Signal Word: Danger

EINECSNumber : 233-245-9