Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-3756653
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
10099-74-8
N2O6Pb
331.20
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
10099-74-8
N2O6Pb
331.20
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 470.0°C
Color White
Physical Form Crystalline Solid
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula Pb(NO3)2
Merck Index 15, 5469
Solubility Information Solubility in water: 343g/L.
Formula Weight 331.2
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.53
Density 4.5300g/mL
Chemical Name or Material Lead(II) nitrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume
EINECSNumber 233-245-9

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit